Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000267.3 | NP_000258.1 | 49 - 50 | 7775 | 134 |
NM_001042492.2 | NP_001035957.1 | 50 - 51 | 7838 | 134 | |
XM_005257983.2 | XP_005258040.1 | 50 - 51 | 7788 | 134 | |
XM_005257984.2 | XP_005258041.1 | 49 - 50 | 7725 | 134 | |
XM_006721922.2 | XP_006721985.1 | 51 - 52 | 7818 | 134 | |
XM_006721924.2 | XP_006721987.1 | 51 - 52 | 7818 | 134 | |
XM_006721925.2 | XP_006721988.1 | 50 - 51 | 7755 | 134 | |
XM_006721926.3 | XP_006721989.1 | 51 - 52 | 7818 | 134 | |
XM_011524852.2 | XP_011523154.1 | 51 - 52 | 7815 | 134 | |
XM_011524857.2 | XP_011523159.1 | 51 - 52 | 7818 | 134 | |
XM_017024689.1 | XP_016880178.1 | 50 - 51 | 7755 | 134 | |
GenBank mRNA | AK301970.1 | - | 7 - 8 | 838 | 134 |
BC144643.1 | - | 50 - 51 | 7591 | 134 | |
M38106.1 | - | 41 - 42 | 6393 | 134 | |
M38107.1 | - | 41 - 42 | 6393 | 134 | |
M60496.1 | - | 7 - 8 | 768 | 134 | |
M82814.1 | - | 49 - 50 | 7603 | 134 | |
M89914.1 | - | 50 - 51 | 7652 | 134 |