Assay Design



Gene Transcripts

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
Chromosome Location
Chr.17: 31094927 - 31377677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000267.3 NP_000258.1 6 - 7 1043 122
NM_001042492.2 NP_001035957.1 6 - 7 1043 122
NM_001128147.2 NP_001121619.1 6 - 7 1043 122
XM_005257983.2 XP_005258040.1 6 - 7 993 122
XM_005257984.2 XP_005258041.1 6 - 7 993 122
XM_006721922.2 XP_006721985.1 6 - 7 993 122
XM_006721924.2 XP_006721987.1 6 - 7 993 122
XM_006721925.2 XP_006721988.1 6 - 7 993 122
XM_006721926.3 XP_006721989.1 6 - 7 993 122
XM_011524852.2 XP_011523154.1 6 - 7 993 122
XM_011524857.2 XP_011523159.1 6 - 7 993 122
XM_017024689.1 XP_016880178.1 6 - 7 993 122
XM_017024690.1 XP_016880179.1 6 - 7 993 122
GenBank mRNA AK289936.1 - 6 - 7 993 122
BC144643.1 - 6 - 7 762 122
BX391799.2 - 3 - 4 122
BX537850.1 - 4 - 5 435 122
D12625.1 - 6 - 7 1043 122
D42072.1 - 6 - 7 725 122
M82814.1 - 6 - 7 871 122
M89914.1 - 6 - 7 857 122

View Full Product Details