Assay Design



Gene Transcripts

Gene Symbol
CCDC12
Entrez Gene ID
151903
Gene Name
coiled-coil domain containing 12
Gene Aliases
-
Chromosome Location
Chr.3: 46917905 - 46982031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_102269.1 1 - 2 147 70
XM_011533390.2 XP_011531692.1 1 - 2 224 70
GenBank mRNA BM803652.1 - 1 - 2 54 70
DA952452.1 - - 147 70

View Full Product Details