Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318798.1 | NP_001305727.1 | 5 - 6 | 491 | 66 |
NM_001318800.1 | NP_001305729.1 | 5 - 6 | 775 | 66 | |
XM_005265382.4 | XP_005265439.1 | 5 - 6 | 592 | 66 | |
GenBank mRNA | AB209259.1 | - | 5 - 6 | 445 | 66 |
BQ949723.1 | - | 5 - 6 | 600 | 66 | |
U38276.1 | - | 5 - 6 | 772 | 66 |