Assay Design



Gene Transcripts

Gene Symbol
POMGNT2
Entrez Gene ID
84892
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-)
Gene Aliases
AGO61, C3orf39, GTDC2, MDDGA8
Chromosome Location
Chr.3: 43079229 - 43106092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032806.5 NP_116195.2 2 - 2 2381 124
XM_005265515.2 XP_005265572.1 3 - 3 2512 124
XM_011534163.2 XP_011532465.1 3 - 3 2500 124
XM_011534164.1 XP_011532466.1 2 - 2 3082 124
XM_017007353.1 XP_016862842.1 4 - 4 2591 124
GenBank mRNA AK027472.1 - 1 - 1 1641 124
AK092147.1 - 2 - 2 2169 124
AK124737.1 - 1 - 1 3349 124
BC060861.1 - 2 - 2 2371 124

View Full Product Details