Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015527.3 | NP_056342.3 | 6 - 7 | 1468 | 92 |
XM_011545789.1 | XP_011544091.1 | 3 - 4 | 1210 | 92 | |
GenBank mRNA | AB449895.1 | - | 6 - 7 | 985 | 92 |
AF370370.1 | - | 6 - 7 | 614 | 92 | |
AK023192.1 | - | 6 - 7 | 847 | 92 | |
BC050523.2 | - | 2 - 3 | 182 | 92 | |
BC063112.1 | - | 6 - 7 | 892 | 92 | |
BC072453.1 | - | 6 - 7 | 780 | 92 | |
BC093814.1 | - | 6 - 7 | 696 | 92 | |
BC093816.1 | - | 6 - 7 | 696 | 92 | |
BC098419.1 | - | 6 - 7 | 1032 | 92 |