Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143981.1 | NP_001137453.1 | 9 - 10 | 1336 | 76 |
NM_001143982.1 | NP_001137454.1 | 9 - 10 | 1333 | 76 | |
NM_001143983.2 | NP_001137455.2 | 7 - 8 | 1096 | 76 | |
XM_005262221.1 | XP_005262278.1 | 9 - 10 | 1153 | 76 | |
XM_005262222.3 | XP_005262279.1 | 9 - 10 | 1150 | 76 | |
GenBank mRNA | AB209548.1 | - | 8 - 9 | 926 | 76 |
AK092245.1 | - | 6 - 7 | 763 | 76 | |
AK293106.1 | - | 9 - 10 | 1083 | 76 | |
AK297563.1 | - | 7 - 8 | 857 | 76 | |
AK312270.1 | - | 9 - 10 | 1094 | 76 | |
AY608914.1 | - | 8 - 9 | 1010 | 76 |