Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000555.3 | NP_000546.2 | 6 - 7 | 1442 | 85 |
NM_178151.2 | NP_835364.1 | 6 - 7 | 1271 | 85 | |
NM_178152.2 | NP_835365.1 | 6 - 7 | 1159 | 85 | |
NM_178153.2 | NP_835366.1 | 6 - 7 | 1144 | 85 | |
GenBank mRNA | AF034634.1 | - | 5 - 6 | 1027 | 85 |
AF040254.1 | - | 6 - 7 | 1084 | 85 | |
AF040255.1 | - | 6 - 7 | 1069 | 85 | |
AJ003112.1 | - | 6 - 7 | 1442 | 85 | |
AK297297.1 | - | 6 - 7 | 1260 | 85 | |
BC027925.1 | - | 6 - 7 | 1086 | 85 |