Assay Design



Gene Transcripts

Gene Symbol
PHF3
Entrez Gene ID
23469
Gene Name
PHD finger protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 63635802 - 63715522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290259.1 NP_001277188.1 11 - 12 3800 76
NM_015153.3 NP_055968.1 9 - 10 3259 76
XM_005248701.3 XP_005248758.1 10 - 11 3594 76
XM_005248702.3 XP_005248759.1 9 - 10 3338 76
XM_005248703.3 XP_005248760.1 8 - 9 1548 76
XM_006715426.3 XP_006715489.1 10 - 11 3819 76
XM_011535648.2 XP_011533950.1 10 - 11 3476 76
XM_017010626.1 XP_016866115.1 9 - 10 1811 76
GenBank mRNA AF091622.1 - 9 - 10 3260 76
AK298580.1 - 8 - 9 1532 76
AK307869.1 - 8 - 9 3159 76
BC113650.1 - 9 - 10 3260 76
BC113652.1 - 9 - 10 3260 76
BX648268.1 - 11 - 12 3486 76
D87685.2 - 9 - 10 3248 76

View Full Product Details