Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014936.4 | NP_055751.1 | 2 - 3 | 1081 | 144 |
GenBank mRNA | AB020686.1 | - | 1 - 2 | 870 | 144 |
AK291276.1 | - | 2 - 3 | 926 | 144 | |
BC018054.1 | - | 2 - 3 | 1051 | 144 |