Assay Design
Gene Transcripts
- Gene Symbol
- RAD52
- Entrez Gene ID
-
5893
- Gene Name
- RAD52 homolog, DNA repair protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 911028 - 991195 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF125948.1 |
- |
7 - 8 |
991 |
75 |
View Full Product Details