Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105192.2 | NP_001098662.1 | 12 - 13 | 2176 | 69 |
NM_001282979.1 | NP_001269908.1 | 12 - 13 | 2176 | 69 | |
NM_001282981.1 | NP_001269910.1 | 12 - 13 | 1109 | 69 | |
XM_005254622.4 | XP_005254679.1 | 12 - 13 | 2224 | 69 | |
XM_005254623.3 | XP_005254680.1 | 12 - 13 | 2224 | 69 | |
XM_006720665.3 | XP_006720728.1 | 12 - 13 | 2191 | 69 | |
GenBank mRNA | AK304459.1 | - | 3 - 4 | 324 | 69 |
AK315058.1 | - | 13 - 14 | 1504 | 69 | |
AY583459.1 | - | 12 - 13 | 1057 | 69 |