Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282979.1 | NP_001269908.1 | 13 - 14 | 2357 | 132 |
NM_001282980.1 | NP_001269909.1 | 13 - 14 | 2366 | 132 | |
NM_001282982.1 | NP_001269911.1 | 11 - 12 | 1112 | 132 | |
XM_005254623.3 | XP_005254680.1 | 13 - 14 | 2405 | 132 | |
XM_005254628.3 | XP_005254685.1 | 13 - 14 | 2378 | 132 | |
XM_005254633.2 | XP_005254690.1 | 13 - 14 | 2348 | 132 | |
XM_006720665.3 | XP_006720728.1 | 13 - 14 | 2372 | 132 | |
XM_011521978.2 | XP_011520280.1 | 13 - 14 | 2414 | 132 | |
XM_011521979.2 | XP_011520281.1 | 12 - 13 | 1220 | 132 | |
XM_011521983.2 | XP_011520285.1 | 11 - 12 | 2162 | 132 | |
GenBank mRNA | AB209672.1 | - | 1 - 2 | 1319 | 132 |
AK001258.1 | - | 2 - 3 | 208 | 132 | |
AK096779.1 | - | 11 - 12 | 1112 | 132 | |
AY583459.1 | - | 13 - 14 | 1238 | 132 | |
AY583460.1 | - | 13 - 14 | 1247 | 132 | |
BC041831.1 | - | 12 - 13 | 1563 | 132 | |
BC043247.2 | - | 13 - 14 | 2365 | 132 |