Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000436.3 | NP_000427.1 | 3 - 4 | 609 | 115 |
XM_017009494.1 | XP_016864983.1 | 3 - 4 | 490 | 115 | |
GenBank mRNA | AK312327.1 | - | 3 - 4 | 115 | |
BC009001.1 | - | 3 - 4 | 333 | 115 | |
CR457049.1 | - | 3 - 4 | 278 | 115 | |
U62961.1 | - | 3 - 4 | 376 | 115 |