Assay Design



Gene Transcripts

Gene Symbol
PRDM2
Entrez Gene ID
7799
Gene Name
PR domain 2
Gene Aliases
HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2
Chromosome Location
Chr.1: 13698875 - 13825079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012231.4 NP_036363.2 8 - 9 5896 68
XM_017002253.1 XP_016857742.1 7 - 8 5118 68
XM_017002255.1 XP_016857744.1 7 - 8 5128 68
XM_017002256.1 XP_016857745.1 8 - 9 5197 68
XM_017002258.1 XP_016857747.1 5 - 6 4828 68
XM_017002259.1 XP_016857748.1 4 - 5 4657 68
XM_017002260.1 XP_016857749.1 3 - 4 5023 68
XM_017002261.1 XP_016857750.1 3 - 4 4738 68
XM_017002263.1 XP_016857752.1 3 - 4 4551 68
GenBank mRNA AB208877.1 - 3 - 4 4615 68
BF378912.1 - 1 - 2 286 68
BM008289.1 - 1 - 2 307 68
U17838.1 - 8 - 9 5898 68

View Full Product Details