Assay Design



Gene Transcripts

Gene Symbol
MTSS1
Entrez Gene ID
9788
Gene Name
metastasis suppressor 1
Gene Aliases
MIM, MIMA, MIMB
Chromosome Location
Chr.8: 124550770 - 124728507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282971.1 NP_001269900.1 4 - 5 846 83
NM_001282974.1 NP_001269903.1 4 - 5 846 83
NM_014751.5 NP_055566.3 4 - 5 846 83
XM_005251111.1 XP_005251168.1 4 - 5 812 83
XM_005251113.1 XP_005251170.1 4 - 5 812 83
XM_005251118.1 XP_005251175.1 4 - 5 812 83
XM_006716700.1 XP_006716763.1 4 - 5 812 83
XM_006716701.1 XP_006716764.1 4 - 5 812 83
XM_006716702.1 XP_006716765.1 4 - 5 812 83
XM_006716703.1 XP_006716766.1 4 - 5 812 83
XM_006716704.1 XP_006716767.1 4 - 5 812 83
XM_006716705.1 XP_006716768.1 4 - 5 812 83
XM_006716706.1 XP_006716769.1 4 - 5 812 83
XM_011517403.1 XP_011515705.1 4 - 5 278 83
XM_011517404.2 XP_011515706.1 2 - 3 11978 83
XM_017014086.1 XP_016869575.1 4 - 5 812 83
XM_017014087.1 XP_016869576.1 4 - 5 812 83
XM_017014088.1 XP_016869577.1 4 - 5 812 83
XM_017014089.1 XP_016869578.1 4 - 5 812 83
XM_017014090.1 XP_016869579.1 2 - 3 1358 83
XM_017014091.1 XP_016869580.1 4 - 5 246 83
XM_017014092.1 XP_016869581.1 4 - 5 812 83
XM_017014093.1 XP_016869582.1 4 - 5 812 83
XM_017014094.1 XP_016869583.1 4 - 5 812 83
XM_017014095.1 XP_016869584.1 4 - 5 812 83
XM_017014096.1 XP_016869585.1 4 - 5 812 83
XM_017014097.1 XP_016869586.1 4 - 5 812 83
XM_017014098.1 XP_016869587.1 4 - 5 812 83
XM_017014099.1 XP_016869588.1 4 - 5 812 83
XM_017014100.1 XP_016869589.1 4 - 5 812 83
XM_017014101.1 XP_016869590.1 2 - 3 471 83
GenBank mRNA AB007889.2 - 1 - 2 103 83
AF086645.1 - 4 - 5 757 83
AK027015.1 - 4 - 5 612 83
EF560715.1 - 4 - 5 477 83

View Full Product Details