Assay Design
Gene Transcripts
- Gene Symbol
- BCR
- Entrez Gene ID
-
613
- Gene Name
- BCR, RhoGEF and GTPase activating protein
- Gene Aliases
-
ALL, BCR1, CML, D22S11, D22S662, PHL
- Chromosome Location
-
Chr.22: 23180365 - 23318037 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X14677.1 |
- |
1 - 2 |
134 |
122 |
View Full Product Details