Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320851.1 | NP_001307780.1 | 1 - 2 | 82 | 78 |
XM_017020818.1 | XP_016876307.1 | 1 - 2 | 76 | 78 | |
XM_017020821.1 | XP_016876310.1 | 1 - 2 | 34 | 78 | |
XM_017020822.1 | XP_016876311.1 | 1 - 2 | 115 | 78 | |
XM_017020824.1 | XP_016876313.1 | 1 - 2 | 76 | 78 | |
XM_017020826.1 | XP_016876315.1 | 1 - 2 | 76 | 78 | |
XM_017020827.1 | XP_016876316.1 | 1 - 2 | 116 | 78 | |
GenBank mRNA | AK300157.1 | - | 1 - 2 | 40 | 78 |
DA404477.1 | - | 1 - 2 | 76 | 78 | |
DA439385.1 | - | 1 - 2 | 82 | 78 |