Assay Design



Gene Transcripts

Gene Symbol
NFIB
Entrez Gene ID
4781
Gene Name
nuclear factor I B
Gene Aliases
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3
Chromosome Location
Chr.9: 14081843 - 14398983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005251470.2 XP_005251527.1 8 - 9 1868 118
XM_017014742.1 XP_016870231.1 7 - 8 3229 118
GenBank mRNA AK299605.1 - 8 - 9 1638 118

View Full Product Details