Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002736.2 | NP_002727.2 | 5 - 6 | 760 | 153 |
GenBank mRNA | AK128639.1 | - | 6 - 7 | 570 | 153 |
AK291441.1 | - | 5 - 6 | 611 | 153 | |
AL559918.3 | - | 5 - 6 | 838 | 153 | |
BC075800.1 | - | 5 - 6 | 799 | 153 | |
M31158.1 | - | 5 - 6 | 751 | 153 |