Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002736.2 | NP_002727.2 | 7 - 8 | 1017 | 142 |
XM_011516398.1 | XP_011514700.1 | 6 - 7 | 938 | 142 | |
GenBank mRNA | AK128639.1 | - | 8 - 9 | 827 | 142 |
AK291441.1 | - | 7 - 8 | 868 | 142 | |
BC075800.1 | - | 7 - 8 | 1056 | 142 | |
M31158.1 | - | 7 - 8 | 1008 | 142 |