Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007235.4 | NP_009166.2 | 22 - 23 | 3239 | 93 |
XM_017018748.1 | XP_016874237.1 | 22 - 23 | 3099 | 93 | |
GenBank mRNA | AF039022.1 | - | 21 - 22 | 2733 | 93 |
AK292884.1 | - | 22 - 23 | 3052 | 93 | |
BC000950.2 | - | 6 - 7 | 957 | 93 | |
BC020569.1 | - | 21 - 22 | 2818 | 93 | |
Y16414.1 | - | 21 - 22 | 2791 | 93 |