Assay Design
Gene Transcripts
- Gene Symbol
- XPOT
- Entrez Gene ID
-
11260
- Gene Name
- exportin for tRNA
- Gene Aliases
-
XPO3
- Chromosome Location
-
Chr.12: 64404350 - 64451125 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK308108.1 |
- |
4 - 5 |
529 |
126 |
View Full Product Details