Assay Design



Gene Transcripts

Gene Symbol
UNG
Entrez Gene ID
7374
Gene Name
uracil DNA glycosylase
Gene Aliases
DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2
Chromosome Location
Chr.12: 109097594 - 109110993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080911.2 NP_550433.1 1 - 2 218 142
GenBank mRNA AK313552.1 - 1 - 2 215 142
BQ950839.1 - 1 - 2 229 142
Y09008.1 - 1 - 2 202 142

View Full Product Details