Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302084.1 | NP_001289013.1 | 13 - 14 | 1464 | 78 |
NM_024650.3 | NP_078926.3 | 15 - 16 | 1756 | 78 | |
GenBank mRNA | AK026184.1 | - | 13 - 14 | 1446 | 78 |
AK225435.1 | - | 13 - 14 | 1446 | 78 | |
AK302025.1 | - | 13 - 14 | 1416 | 78 | |
BC028240.1 | - | 15 - 16 | 1759 | 78 |