Assay Design



Gene Transcripts

Gene Symbol
OSBPL1A
Entrez Gene ID
114876
Gene Name
oxysterol binding protein like 1A
Gene Aliases
ORP-1, ORP1, OSBPL1B
Chromosome Location
Chr.18: 24162045 - 24397882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080597.3 NP_542164.2 10 - 11 1025 79
XM_006722380.3 XP_006722443.1 9 - 10 938 79
XM_006722382.3 XP_006722445.1 10 - 11 1038 79
XM_017025530.1 XP_016881019.1 10 - 11 1317 79
XM_017025531.1 XP_016881020.1 9 - 10 810 79
XM_017025532.1 XP_016881021.1 9 - 10 810 79
GenBank mRNA AF323726.2 - 10 - 11 982 79
AF392450.1 - 10 - 11 982 79
AK096358.1 - 10 - 11 1025 79
AK292895.1 - 10 - 11 957 79
BC136452.1 - 10 - 11 922 79
BC144117.1 - 10 - 11 922 79
BX647893.1 - 4 - 5 2699 79

View Full Product Details