Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002298.4 | NP_002289.2 | 6 - 7 | 814 | 91 |
XM_005266374.1 | XP_005266431.1 | 6 - 7 | 712 | 91 | |
GenBank mRNA | AB208984.1 | - | 6 - 7 | 659 | 91 |
AK223305.1 | - | 6 - 7 | 749 | 91 | |
AK312393.1 | - | 6 - 7 | 680 | 91 | |
BC007673.1 | - | 6 - 7 | 652 | 91 | |
BC010271.1 | - | 6 - 7 | 716 | 91 | |
CR936795.1 | - | 6 - 7 | 702 | 91 | |
EU794648.1 | - | 6 - 7 | 652 | 91 | |
J02923.1 | - | 6 - 7 | 650 | 91 | |
M22300.1 | - | 4 - 5 | 405 | 91 |