Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013336.3 | NP_037468.1 | 4 - 5 | 404 | 90 |
GenBank mRNA | AF077032.1 | - | 4 - 5 | 310 | 90 |
AF084458.1 | - | 4 - 5 | 310 | 90 | |
AF346602.1 | - | 4 - 5 | 286 | 90 | |
AK027423.1 | - | 4 - 5 | 404 | 90 | |
AK074706.1 | - | 4 - 5 | 404 | 90 | |
AK299118.1 | - | 4 - 5 | 484 | 90 | |
AL832821.1 | - | 4 - 5 | 337 | 90 | |
BC002951.1 | - | 4 - 5 | 311 | 90 |