Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013336.3 | NP_037468.1 | 9 - 10 | 1158 | 90 |
GenBank mRNA | AF084458.1 | - | 9 - 10 | 1064 | 90 |
AF346602.1 | - | 9 - 10 | 1040 | 90 | |
AK001689.1 | - | 2 - 3 | 218 | 90 | |
AK027423.1 | - | 9 - 10 | 1158 | 90 | |
AK027452.1 | - | 1 - 2 | 124 | 90 | |
AK074706.1 | - | 9 - 10 | 1158 | 90 | |
AK074907.1 | - | 1 - 2 | 128 | 90 | |
AK074928.1 | - | 5 - 6 | 766 | 90 | |
AK075148.1 | - | 1 - 2 | 158 | 90 | |
AK299118.1 | - | 9 - 10 | 1238 | 90 | |
AL831940.1 | - | 4 - 5 | 568 | 90 | |
AL832821.1 | - | 9 - 10 | 1091 | 90 | |
BC002951.1 | - | 9 - 10 | 1065 | 90 |