Assay Design



Gene Transcripts

Gene Symbol
CCDC6
Entrez Gene ID
8030
Gene Name
coiled-coil domain containing 6
Gene Aliases
D10S170, H4, PTC, TPC, TST1
Chromosome Location
Chr.10: 59788747 - 59906656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005436.4 NP_005427.2 3 - 4 817 140
GenBank mRNA AK292593.1 - 3 - 4 817 140
BC022284.1 - 3 - 4 711 140
BC036757.2 - 3 - 4 656 140
BC064391.1 - 3 - 4 643 140
S72869.1 - 3 - 4 621 140

View Full Product Details