Assay Design



Gene Transcripts

Gene Symbol
KCNK10
Entrez Gene ID
54207
Gene Name
potassium two pore domain channel subfamily K member 10
Gene Aliases
K2p10.1, PPP1R97, TREK-2, TREK2
Chromosome Location
Chr.14: 88180108 - 88326912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138318.2 NP_612191.1 1 - 2 176 77
GenBank mRNA AF385399.1 - 1 - 2 177 77
BC075021.2 - 1 - 2 95 77
BC075022.2 - 1 - 2 92 77
EU978940.1 - 1 - 2 82 77
EU978941.1 - 1 - 2 82 77

View Full Product Details