Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004562.2 | NP_004553.2 | 1 - 2 | 142 | 76 |
NM_013987.2 | NP_054642.2 | 1 - 2 | 142 | 76 | |
NM_013988.2 | NP_054643.2 | 1 - 2 | 142 | 76 | |
XM_011535863.1 | XP_011534165.1 | 1 - 2 | 142 | 76 | |
GenBank mRNA | AB009973.1 | - | 1 - 2 | 109 | 76 |
AB245403.1 | - | 1 - 2 | 100 | 76 | |
AK292590.1 | - | 1 - 2 | 103 | 76 | |
BC022014.2 | - | 1 - 2 | 106 | 76 | |
DB023187.1 | - | 1 - 2 | 142 | 76 |