Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 11 - 12 | 1272 | 145 |
NM_020320.4 | NP_064716.2 | 12 - 13 | 1107 | 145 | |
XM_011535948.2 | XP_011534250.1 | 12 - 13 | 1092 | 145 | |
XM_011535949.2 | XP_011534251.1 | 12 - 13 | 1092 | 145 | |
XM_017011072.1 | XP_016866561.1 | 12 - 13 | 1273 | 145 | |
XM_017011073.1 | XP_016866562.1 | 13 - 14 | 1491 | 145 | |
XM_017011074.1 | XP_016866563.1 | 12 - 13 | 1094 | 145 | |
XM_017011075.1 | XP_016866564.1 | 11 - 12 | 1217 | 145 | |
XM_017011076.1 | XP_016866565.1 | 13 - 14 | 1435 | 145 | |
XM_017011077.1 | XP_016866566.1 | 11 - 12 | 994 | 145 | |
XM_017011078.1 | XP_016866567.1 | 11 - 12 | 1038 | 145 | |
XM_017011079.1 | XP_016866568.1 | 12 - 13 | 1273 | 145 | |
XM_017011080.1 | XP_016866569.1 | 13 - 14 | 1435 | 145 | |
XM_017011081.1 | XP_016866570.1 | 11 - 12 | 1038 | 145 | |
GenBank mRNA | AK023550.1 | - | 12 - 13 | 1050 | 145 |
AK093934.1 | - | 12 - 13 | 1480 | 145 | |
AK315669.1 | - | 12 - 13 | 1044 | 145 | |
BC010420.1 | - | 12 - 13 | 1078 | 145 | |
BC105784.1 | - | 11 - 12 | 1211 | 145 |