Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 4 - 5 | 600 | 139 |
NM_020320.4 | NP_064716.2 | 4 - 5 | 379 | 139 | |
NR_134857.1 | 4 - 5 | 379 | 139 | ||
XM_011535948.2 | XP_011534250.1 | 4 - 5 | 364 | 139 | |
XM_011535949.2 | XP_011534251.1 | 4 - 5 | 364 | 139 | |
XM_017011072.1 | XP_016866561.1 | 4 - 5 | 545 | 139 | |
XM_017011073.1 | XP_016866562.1 | 5 - 6 | 763 | 139 | |
XM_017011074.1 | XP_016866563.1 | 4 - 5 | 366 | 139 | |
XM_017011075.1 | XP_016866564.1 | 4 - 5 | 545 | 139 | |
XM_017011076.1 | XP_016866565.1 | 4 - 5 | 545 | 139 | |
XM_017011077.1 | XP_016866566.1 | 4 - 5 | 322 | 139 | |
XM_017011078.1 | XP_016866567.1 | 4 - 5 | 366 | 139 | |
XM_017011079.1 | XP_016866568.1 | 4 - 5 | 545 | 139 | |
XM_017011080.1 | XP_016866569.1 | 4 - 5 | 545 | 139 | |
XM_017011081.1 | XP_016866570.1 | 4 - 5 | 366 | 139 | |
GenBank mRNA | AK023550.1 | - | 4 - 5 | 322 | 139 |
AK093934.1 | - | 4 - 5 | 752 | 139 | |
AK315669.1 | - | 4 - 5 | 316 | 139 | |
BC010420.1 | - | 4 - 5 | 350 | 139 | |
BC105784.1 | - | 4 - 5 | 539 | 139 | |
DA607375.1 | - | 4 - 5 | 365 | 139 | |
DB455377.1 | - | 4 - 5 | 381 | 139 |