Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020320.4 | NP_064716.2 | 6 - 7 | 526 | 158 |
NR_134857.1 | 6 - 7 | 526 | 158 | ||
XM_011535948.2 | XP_011534250.1 | 6 - 7 | 511 | 158 | |
XM_011535949.2 | XP_011534251.1 | 6 - 7 | 511 | 158 | |
XM_017011072.1 | XP_016866561.1 | 6 - 7 | 692 | 158 | |
XM_017011073.1 | XP_016866562.1 | 7 - 8 | 910 | 158 | |
XM_017011074.1 | XP_016866563.1 | 6 - 7 | 513 | 158 | |
XM_017011079.1 | XP_016866568.1 | 6 - 7 | 692 | 158 | |
GenBank mRNA | AK093934.1 | - | 6 - 7 | 899 | 158 |
AK315669.1 | - | 6 - 7 | 463 | 158 | |
BC010420.1 | - | 6 - 7 | 497 | 158 |