Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001441.2 | NP_001432.2 | 4 - 5 | 659 | 69 |
GenBank mRNA | BC093632.1 | - | 4 - 5 | 601 | 69 |
BC110404.1 | - | 4 - 5 | 589 | 69 | |
BC111941.1 | - | 4 - 5 | 601 | 69 | |
BG718115.1 | - | 4 - 5 | 666 | 68 | |
U82535.1 | - | 4 - 5 | 612 | 69 |