Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052875.4 | NP_443107.1 | 1 - 2 | 738 | 59 |
XM_011542565.2 | XP_011540867.1 | 1 - 2 | 714 | 59 | |
GenBank mRNA | AF452718.1 | - | 1 - 2 | 676 | 59 |
BC009747.2 | - | 1 - 2 | 654 | 59 | |
BC014128.2 | - | 1 - 2 | 663 | 59 | |
BC098386.1 | - | 1 - 2 | 739 | 59 |