Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020911.1 | NP_065962.1 | 17 - 18 | 3476 | 70 |
XM_005250686.4 | XP_005250743.1 | 17 - 18 | 3874 | 70 | |
XM_006716171.3 | XP_006716234.1 | 18 - 19 | 3967 | 70 | |
XM_011516676.2 | XP_011514978.1 | 17 - 18 | 3382 | 70 | |
XM_017012779.1 | XP_016868268.1 | 16 - 17 | 3181 | 70 | |
GenBank mRNA | AB046770.3 | - | 15 - 16 | 1953 | 70 |