Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020911.1 | NP_065962.1 | 9 - 10 | 2326 | 71 |
XM_005250686.4 | XP_005250743.1 | 9 - 10 | 2724 | 71 | |
XM_006716171.3 | XP_006716234.1 | 10 - 11 | 2817 | 71 | |
XM_011516676.2 | XP_011514978.1 | 9 - 10 | 2232 | 71 | |
GenBank mRNA | AB046770.3 | - | 7 - 8 | 803 | 71 |