Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025247.1 | NP_001020418.1 | 1 - 2 | 164 | 96 |
NM_014409.3 | NP_055224.1 | 1 - 2 | 164 | 96 | |
XM_011544163.1 | XP_011542465.1 | 1 - 2 | 150 | 96 | |
GenBank mRNA | AI251746.1 | - | 1 - 2 | 164 | 96 |
BC041094.1 | - | 1 - 2 | 87 | 96 | |
BE646579.1 | - | 1 - 2 | 155 | 96 |