Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005689.2 | NP_005680.1 | 5 - 6 | 1476 | 94 |
GenBank mRNA | AF070598.1 | - | 1 - 2 | 168 | 94 |
AF076775.1 | - | 5 - 6 | 1159 | 94 | |
AF308472.1 | - | 5 - 6 | 1465 | 94 | |
AJ289233.2 | - | 5 - 6 | 1436 | 94 | |
AK026067.1 | - | 2 - 3 | 285 | 94 | |
AK057026.1 | - | 5 - 6 | 1195 | 94 | |
AK172812.1 | - | 5 - 6 | 826 | 94 | |
BC000559.2 | - | 5 - 6 | 1427 | 94 |