Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005689.2 | NP_005680.1 | 8 - 9 | 1769 | 61 |
GenBank mRNA | AF070598.1 | - | 4 - 5 | 461 | 61 |
AF076775.1 | - | 8 - 9 | 1452 | 61 | |
AF308472.1 | - | 8 - 9 | 1758 | 61 | |
AJ289233.2 | - | 8 - 9 | 1729 | 61 | |
AK026067.1 | - | 5 - 6 | 578 | 61 | |
AK057026.1 | - | 8 - 9 | 1488 | 61 | |
AK172812.1 | - | 8 - 9 | 1119 | 61 | |
BC000559.2 | - | 8 - 9 | 1720 | 61 | |
BC043423.1 | - | 1 - 2 | 1769 | 61 |