Assay Design
Gene Transcripts
- Gene Symbol
- ABCB6
- Entrez Gene ID
-
10058
- Gene Name
- ATP binding cassette subfamily B member 6 (Langereis blood group)
- Gene Aliases
-
ABC, ABC14, DUH3, LAN, MCOPCB7, MTABC3, PRP, PSHK2, umat
- Chromosome Location
-
Chr.2: 219209766 - 219218990 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK303230.1 |
- |
3 - 4 |
907 |
66 |
View Full Product Details