Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300960.1 | NP_001287889.1 | 3 - 4 | 490 | 71 |
NM_001300963.1 | NP_001287892.1 | 4 - 5 | 306 | 71 | |
NM_001300964.1 | NP_001287893.1 | 3 - 4 | 260 | 71 | |
NM_015076.4 | NP_055891.1 | 3 - 4 | 490 | 71 | |
XM_011535630.2 | XP_011533932.1 | 4 - 5 | 968 | 71 | |
XM_011535632.1 | XP_011533934.1 | 2 - 3 | 182 | 71 | |
XM_017010587.1 | XP_016866076.1 | 4 - 5 | 716 | 71 | |
XM_017010588.1 | XP_016866077.1 | 2 - 3 | 240 | 71 | |
GenBank mRNA | AB028951.2 | - | 3 - 4 | 307 | 71 |
AK292360.1 | - | 3 - 4 | 398 | 71 | |
AY028424.1 | - | 3 - 4 | 490 | 71 | |
BC024247.1 | - | 4 - 5 | 311 | 71 | |
BC037289.1 | - | 3 - 4 | 385 | 71 | |
DB030002.1 | - | 3 - 4 | 262 | 71 | |
JX028238.1 | - | 3 - 4 | 310 | 71 |