Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300963.1 | NP_001287892.1 | 7 - 8 | 636 | 152 |
NM_001300964.1 | NP_001287893.1 | 6 - 7 | 590 | 152 | |
NM_015076.4 | NP_055891.1 | 6 - 7 | 820 | 152 | |
XM_005266871.3 | XP_005266928.1 | 3 - 4 | 403 | 152 | |
XM_011535631.2 | XP_011533933.1 | 5 - 6 | 1038 | 152 | |
XM_011535632.1 | XP_011533934.1 | 5 - 6 | 512 | 152 | |
XM_017010587.1 | XP_016866076.1 | 7 - 8 | 1046 | 152 | |
XM_017010588.1 | XP_016866077.1 | 4 - 5 | 512 | 152 | |
GenBank mRNA | AB028951.2 | - | 6 - 7 | 637 | 152 |
AK292360.1 | - | 6 - 7 | 728 | 152 | |
AK300567.1 | - | 3 - 4 | 312 | 152 | |
AL122055.1 | - | 3 - 4 | 275 | 152 | |
AY028424.1 | - | 6 - 7 | 820 | 152 | |
BC024247.1 | - | 7 - 8 | 641 | 152 | |
BC037289.1 | - | 6 - 7 | 715 | 152 |