Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300960.1 | NP_001287889.1 | 8 - 9 | 988 | 74 |
NM_001300963.1 | NP_001287892.1 | 10 - 11 | 936 | 74 | |
NM_001300964.1 | NP_001287893.1 | 9 - 10 | 890 | 74 | |
NM_015076.4 | NP_055891.1 | 9 - 10 | 1120 | 74 | |
XM_005266871.3 | XP_005266928.1 | 6 - 7 | 703 | 74 | |
XM_011535630.2 | XP_011533932.1 | 9 - 10 | 1466 | 74 | |
XM_011535631.2 | XP_011533933.1 | 8 - 9 | 1338 | 74 | |
XM_017010587.1 | XP_016866076.1 | 10 - 11 | 1346 | 74 | |
XM_017010588.1 | XP_016866077.1 | 7 - 8 | 812 | 74 | |
GenBank mRNA | AB028951.2 | - | 9 - 10 | 937 | 74 |
AK292360.1 | - | 9 - 10 | 1028 | 74 | |
AK300567.1 | - | 6 - 7 | 612 | 74 | |
AL122055.1 | - | 6 - 7 | 575 | 74 | |
AY028424.1 | - | 9 - 10 | 1120 | 74 | |
BC024247.1 | - | 10 - 11 | 941 | 74 | |
BC037289.1 | - | 9 - 10 | 1015 | 74 | |
JX028238.1 | - | 8 - 9 | 808 | 74 |