Assay Design



Gene Transcripts

Gene Symbol
GPA33
Entrez Gene ID
10223
Gene Name
glycoprotein A33
Gene Aliases
A33
Chromosome Location
Chr.1: 167052836 - 167090631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005814.2 NP_005805.1 3 - 4 766 52
XM_017000005.1 XP_016855494.1 4 - 5 594 52
GenBank mRNA BC069705.1 - 3 - 4 766 52
BC069723.1 - 3 - 4 766 52
BC069745.1 - 3 - 4 766 52
BC069761.1 - 3 - 4 766 52
BC069789.1 - 3 - 4 766 52
BC074830.2 - 3 - 4 444 52
BC074876.2 - 3 - 4 444 52
BC107164.1 - 3 - 4 721 52
BC107165.1 - 3 - 4 723 52
U79725.1 - 3 - 4 766 52

View Full Product Details