Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144702.2 | NP_653303.2 | 3 - 4 | 609 | 62 |
XM_005244926.3 | XP_005244983.1 | 3 - 4 | 633 | 62 | |
XM_005244927.3 | XP_005244984.1 | 3 - 4 | 633 | 62 | |
XM_005244928.2 | XP_005244985.1 | 3 - 4 | 633 | 62 | |
XM_006711185.3 | XP_006711248.1 | 4 - 5 | 423 | 62 | |
XM_006711186.3 | XP_006711249.1 | 3 - 4 | 633 | 62 | |
XM_006711187.3 | XP_006711250.1 | 3 - 4 | 633 | 62 | |
XM_011509241.2 | XP_011507543.1 | 3 - 4 | 633 | 62 | |
XM_017000459.1 | XP_016855948.1 | 3 - 4 | 633 | 62 | |
XM_017000460.1 | XP_016855949.1 | 3 - 4 | 941 | 62 | |
XM_017000461.1 | XP_016855950.1 | 4 - 5 | 479 | 62 | |
XM_017000462.1 | XP_016855951.1 | 3 - 4 | 365 | 62 | |
GenBank mRNA | AK057446.1 | - | 4 - 5 | 421 | 62 |
AK292342.1 | - | 3 - 4 | 609 | 62 | |
BC033790.1 | - | 3 - 4 | 591 | 62 |