Assay Design



Gene Transcripts

Gene Symbol
ZBTB44
Entrez Gene ID
29068
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
BTBD15, HSPC063, ZNF851
Chromosome Location
Chr.11: 130226677 - 130314908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301098.1 NP_001288027.1 2 - 3 1405 86
NM_001301099.1 NP_001288028.1 2 - 3 1405 86
NM_014155.4 NP_054874.3 2 - 3 1405 86
XM_005271521.4 XP_005271578.1 2 - 3 1601 86
XM_005271522.4 XP_005271579.1 2 - 3 1480 86
XM_006718825.3 XP_006718888.1 2 - 3 1480 86
XM_006718826.3 XP_006718889.1 2 - 3 1477 86
XM_011542789.2 XP_011541091.1 2 - 3 1478 86
XM_017017623.1 XP_016873112.1 2 - 3 1477 86
XM_017017624.1 XP_016873113.1 2 - 3 1477 86
GenBank mRNA AF161548.1 - 2 - 3 1122 86
AK024659.1 - 1 - 2 572 86
BC030580.1 - 2 - 3 1289 86
BC049375.1 - 2 - 3 1310 86
BC050723.1 - 1 - 2 1027 86
BC071729.1 - 2 - 3 1384 86

View Full Product Details