Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005061.2 | NP_005052.1 | 8 - 9 | 1099 | 60 |
XM_011522571.2 | XP_011520873.1 | 8 - 9 | 1559 | 60 | |
GenBank mRNA | AK313162.1 | - | 8 - 9 | 1111 | 60 |
BC050413.1 | - | 8 - 9 | 1091 | 60 | |
U65581.1 | - | 8 - 9 | 1100 | 60 |